Search Fisher Scientific
| Catalog Number | Length (Metric) | Product Type |
|---|---|---|
| MICSTAH-60WH | 152.4 cm | Fixed Handle |
| MICSTAEH-072WH | 101.6 to 182.8 cm | Extendable Handle |
| MICSTAEH-237BL | 172.7 to 602 cm | Extendable Handle |
| MICSTAEH-183WH | 172.7 to 464.8 cm | Extendable Handle |
| MICSTAEH-036WH | 46 to 91 cm | Extendable Handle |
| MICSTAH-24WH | 60.9 cm | Fixed Handle |
Micronova Plastic ACME Threaded Handles are made from lightweight fiberglass coated in plastic and are available in a range of extendable and fixed length options. The ACME Connect threaded ends attach directly to the plastic (QP series) adapters, or use the SSA-2 ACME Thread Adapters to attach to Quick Disconnect (QD series) mop heads.