ACME Thread Plastic Handle
ACME Thread Plastic Handle
Actual product may vary
Micronova
ACME Thread Plastic Handle
Plastic coated fiberglass, fixed length and extendable handles. Designed with ACME Connect threaded ends for use with the entire range of Micronova adapters, mop heads and tools for simplified and versatile cleaning.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Product Type
MICSTAEH-237BL172.7 to 602 cmExtendable Handle
MICSTAEH-072WH101.6 to 182.8 cmExtendable Handle
MICSTAEH-183WH172.7 to 464.8 cmExtendable Handle
MICSTAEH-036WH46 to 91 cmExtendable Handle
MICSTAH-24WH60.9 cmFixed Handle
MICSTAH-60WH152.4 cmFixed Handle
Catalog number MICSTAEH-237BL
Price (SGD)
446.00
CS
Add to cart
Length (Metric):
172.7 to 602 cm
Product Type:
Extendable Handle
Price (SGD)
446.00
CS
Add to cart
Lightweight, plastic-coated fiberglass handles
Micronova Plastic ACME Threaded Handles are made from lightweight fiberglass coated in plastic and are available in a range of extendable and fixed length options. The ACME Connect threaded ends attach directly to the plastic (QP series) adapters, or use the SSA-2 ACME Thread Adapters to attach to Quick Disconnect (QD series) mop heads.
  • Lightweight, plastic coated fiberglass handles
  • ACME Connect threaded end connection
  • Available fixed length or extendable
  • Fixed lengths range from 12 to 60 inches (30 cm to 1.5 m)
  • Extendable maximum lengths range from 36 in to 11.5 ft (90 cm to 3.5 m)
  • ACME Connect threaded end attaches directly to the plastic (QP series) adapters
  • Attach to (QD series) mop heads via the SSA-2 ACME Thread Adapters
  • Autoclavable
Specifications
Color CodeBlack
Product TypeExtendable Handle
ColorWhite, Black
Length (English)68 to 237 in.
SterilityNon-sterile
Length (Metric)172.7 to 602 cm
AutoclavableAutoclavable
Width (Metric)1.9 cm to 2.5 cm
Width (English)0.75 in. to 1 in.
Resistant ToCommon disinfectants and cleaning solutions
DisposableReusable
Thread StyleUniversal Acme; Black End Cap
Quantity1 Ea.
Care InstructionsStore at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
For Use With (Equipment)QP mop adapters and brushes with threaded connection
For Use With (Application)Mop Handle for Cleaning and Sanitizing Cleanrooms
Certifications/ComplianceISO 9001 Certified Facility Certificate of Conformance
Color-codedYes
MaterialFiberglass, Plastic
Diameter (Metric)1.9 cm to 2.5 cm
Unit SizeCS