Stainless Steel Quick Disconnect SSU Handles
Stainless Steel Quick Disconnect SSU Handles
Actual product may vary
Micronova
Stainless Steel Quick Disconnect SSU Handles
The fundamental electropolished stainless steel handles with quick disconnect attachments that work with all Micronova QD series mop adapters and cleaning tools. Available in a range of fixed and extendible lengths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeLength (English)
MICSSU-9Fixed Handle12 in.
MICSSU-1Extendible Handle, Quick Disconnect Attachment38 to 72 in.
MICSSU-3Extendible Handle, Quick Disconnect Attachment62 to 120 in.
MICSSU-4Fixed Handle, Quick Disconnect Attachment61 in.
FIS19-035-361DFixed Handle51 in.
MICSSU-7Fixed Handle51 in.
Catalog number MICSSU-9
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Fixed Handle
Length (English):
12 in.
Entire handle can be autoclaved
The Micronova Stainless Steel Quick Disconnect SSU Handles are made from 304 electropolished stainless steel with polyketone components that provide strong, hard-wearing all-around connections for efficient and long lasting use. The included Quick Disconnect attachment is designed for use with the QD series mop adapters and cleaning tools.
  • Fundamental handles that work with all Micronova mopping systems
  • Electropolished stainless steel with Polyketone components
  • Strong, hard-wearing connections for long lasting use
  • Available fixed or extendible:
    • Fixed length handles available from 8 in (20 cm) to 5 feet (1.5 m)
    • Extension handles available from 3 feet (1 m) to 10 feet (3 m)
  • For use with all QD series adapters
  • May be used with QP tools and frames with addition of the SSA adapter
  • Good compatibility with disinfectants
  • Autoclavable
Specifications
Product TypeFixed Handle
Length (English)12 in.
SterilityNon-sterile
Length (Metric)30.5 cm
AutoclavableAutoclavable
Resistant ToChemicals, Corrosion
Care InstructionsStore at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
For Use With (Equipment)Mops
DescriptionQuick Disconnect Attachment
For Use With (Application)Mop Handle for Cleaning and Sanitizing Cleanrooms
MaterialElectropolished Stainless Steel (304), Polyketone
Unit SizeEA