ACME Thread Adapters
ACME Thread Adapters
Actual product may vary
ACME Thread Adapters
The two versions of stainless steel ACME adapters connect threaded brush and mop adapters to Quick Disconnect, SSU Series Handles (SSA-3), or connect Quick Disconnect mop adapters to threaded handles (SSA-2).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)
MICSSA-35.75 in.
MICSSA-28 in.
Catalog number MICSSA-3
Price (SGD)
118.00
EA
Add to cart
Length (English):
5.75 in.
Price (SGD)
118.00
EA
Add to cart
The Micronova ACME Thread Adapters serve as the connection between various Micronova Adapters and either Threaded or Quick Disconnect handles. The SSA-2 serves to connect a range of Quick Disconnect mop adapters and cleaning tools with threaded handles, such as STA Series Handles. The SSA-3 serves to connect a range of threaded brushes and mop adapters, such as QPSL series, with the Quick Disconnect stainless steel SSU Series Handles.
  • ACME threaded adapters
  • Stainless steel with polyketone connectors
  • Connects various Adapters to either Threaded or Quick Disconnect handles
  • SSA-2 connects threaded handles to QD mop and tool adapters
  • SSA-3 connects threaded mop and brush adapters to QD SSU Handles
  • Autoclavable
Specifications
Product TypeMale Thread Adapter
Length (English)5.75 in.
SterilityNon-sterile
Length (Metric)14.6 cm
AutoclavableAutoclavable
For Use With (Equipment)Threaded Connector for Handles and QD Mop Adapters and Tools
For Use With (Application)Floors, Walls, Isolators
MaterialElectropolished Stainless Steel (304), Polyketone
Unit SizeEA