Search Fisher Scientific
| Catalog Number | Length (English) |
|---|---|
| MICSSA-2 | 8 in. |
| MICSSA-3 | 5.75 in. |
The Micronova ACME Thread Adapters serve as the connection between various Micronova Adapters and either Threaded or Quick Disconnect handles. The SSA-2 serves to connect a range of Quick Disconnect mop adapters and cleaning tools with threaded handles, such as STA Series Handles. The SSA-3 serves to connect a range of threaded brushes and mop adapters, such as QPSL series, with the Quick Disconnect stainless steel SSU Series Handles.