Stainless Steel Gel Cleaner
Actual product may vary
Micronova
Stainless Steel Gel Cleaner
Stainless steel gel cleanser spray, with advanced no-drip technology, for removing rust, oxidation and residue build-up on stainless steel, metal and many other hard surfaces. Available in a 16 oz. spray bottle.
Have Questions?
Catalog NumberVolume (English)
MICSC1-P16 oz.
Catalog number MICSC1-P
Price (SGD)
213.00
Each
Volume (English):
16 oz.
Price (SGD)
213.00
Each
  • Clings and glides over most surfaces with minimum effort on contact
  • Apply into seams and crevices, when dealing with rust, oxidation and residue build-up to pre-treat before wiping
  • Nonatomized — will not affect air quality
  • Phosphorus-free
  • Biodegradable surfactants
  • No VOCs
  • NSF A7 rating
Specifications
Product TypeGel Detergent
Color-codedNo
FormGel
Handle TypeNone
SterilityNon-sterile
AppearanceClear
ColorClear
For Use With (Application)Gel Cleaner for Stainless Surface
Volume (English)16 oz.
Unit SizeEach
Contents & Storage
Store at ambient temperature. Avoid direct sunlight. Protect from freezing.