SwabTool™ Adapters
SwabTool™ Adapters
Actual product may vary
SwabTool™ Adapters
SwabTool™ Adapters
Micronova
SwabTool™ Adapters
A custom tool designed specifically to hold swabs for environmental monitoring and sampling. Available with optional handles for extension in large tanks and other hard to reach areas.
Have Questions?
Catalog NumberAutoclavable
MICQDSWT-01Autoclavable
Catalog number MICQDSWT-01
Price (SGD)
172.00
EA
Add to cart
Autoclavable:
Autoclavable
Price (SGD)
172.00
EA
Add to cart
The Micronova SwabTool™ Adapter locks swabs in place with a twist knob. Compatible with a range of swabs when used with optional 'straws' to hold those which are smaller or longer. Optional SSU series handles available for monitoring and sampling in large tanks and other hard to reach areas.
  • Swab holder for environmental monitoring and sampling
  • Crafted from electropolished stainless steel with acetal components
  • Compatible with a range of swabs
  • Optional 'straws' available to hold smaller or longer swabs
  • QD (Quick Disconnect) version works with SSU series stainless steel handles
  • QP version features an angle joint and works with STA series threaded plastic handles
  • Compatible with strong disinfectants
  • Autoclavable
Specifications
Product TypeFixed Straight Adapter
ColorWhite
Length (English)6.75 in.
SterilityNon-sterile
Length (Metric)17.15 cm
AutoclavableAutoclavable
Width (Metric)3.18 cm
Width (English)1.25 in.
Resistant ToChemicals
DisposableReusable
Care InstructionsStore at Ambient Temperature
For Use With (Equipment)QPSWT-04, SSU Series Handles
For Use With (Application)Environmental Monitoring with Swabs
MaterialAcetal, Electropolished Stainless Steel
Unit SizeEA