Inspection Mirror with Plastic Bottom Plate
Inspection Mirror with Plastic Bottom Plate
Actual product may vary
Inspection Mirror with Plastic Bottom Plate
An electropolished stainless steel mirror with a swivel joint and handle connection for inspecting hard to reach areas.
Have Questions?
Catalog NumberMaterial
MICQDIST-66PStainless Steel, Polypropylene
Catalog number MICQDIST-66P
Price (SGD)
684.00
EA
Add to cart
Material:
Stainless Steel, Polypropylene
Price (SGD)
684.00
EA
Add to cart
  • A highly polished stainless steel mirror for inspecting hard to reach areas
  • Features a swivel connection allowing it to be attached to fixed and extension handles
  • This allows the operator to run the unit behind equipment and fixtures to ensure all surfaces have been properly cleaned
  • The optional flashlight attachment is ideal for use in tanks and other low light applications
  • QD versions connect with SSU series stainless steel handles
  • Specifications
    MaterialStainless Steel, Polypropylene
    Unit SizeEA