Irradiated IPA Resistant LabMarker
Irradiated IPA Resistant LabMarker
Actual product may vary
Micronova
Irradiated IPA Resistant LabMarker
A fine tipped permanent marker with alcohol-resistant ink that marks on cold or wet surfaces, plastic bags and disposable lab-ware.
Have Questions?
Catalog NumberColor
MICPEN-40IRBlack
Catalog number MICPEN-40IR
Price (SGD)
105.00
Special offer
Ends: 31-Mar-2026
144.00
Save 39.00 (27%)
Pack
Add to cart
Color:
Black
Price (SGD)
105.00
Special offer
Ends: 31-Mar-2026
144.00
Save 39.00 (27%)
Pack
Add to cart
With permanent, quick drying black ink that is resistant to IPA
The Micronova Irradiated IPA Resistant LabMarker features permanent, quick drying, alcohol-resistant ink that marks on a range of surfaces and will not smear on reusable glass, metal or porcelain, but can be scrubbed off. Available gamma irradiated for aseptic areas.
  • Fine-tip, black permanent marker
  • Quick drying, alcohol-resistant ink
  • Marks on cold or wet surfaces, plastic bags, and disposable lab-ware
  • Smear-resistant on glass, metal and porcelain, but can be scrubbed off
  • Available gamma irradiated for aseptic areas
Specifications
MaterialHDPE
ColorBlack
Length (English)5.25 in.
AutoclavableNot Autoclavable
DisposableReusable
For Use With (Application)Cleanroom and Aseptic Environment Documentation and Labeling
Packaging10/Pk.
Product TypeCleanroom Pen
QuantityEa.
SterilitySterile
Tip StyleFine
Unit SizePack