NovaClean™ Floor Cleaner
NovaClean™ Floor Cleaner
Actual product may vary
NovaClean™ Floor Cleaner
A concentrated, all-purpose cleanroom detergent that is designed for use between disinfectant applications and for residue removal. Free of mobile ions, this formula is effective on most surfaces, including conductive flooring and semi conductor applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (English)
MICNC1-G1 gal.
FKS119524751 gal.
Catalog number MICNC1-G
Price (SGD)
510.00
Special offer
726.00
Save 216.00 (30%)
CS
Add to cart
Volume (English):
1 gal.
Price (SGD)
510.00
Special offer
726.00
Save 216.00 (30%)
CS
Add to cart
The Micronova NovaClean™ Floor Cleaner is an all-purpose cleanroom detergent with a fine filtration and purity that is ideal for pharmaceutical and biotech industries. NovaClean AFX (NC10-G) is an amine free formulation, particularly suited in semi conductor applications.
  • Concentrated cleanroom detergent
  • Effective on most surfaces, including conductive and semi conductor applications
  • Ideal for pharmaceutical and biotech applications
  • Will not generate a static charge and will not affect the efficiency of conductive flooring
  • Yields 60-128 gallons of cleaning solution per 1 gallon container
  • Free of mobile ions
  • Filtered to 0.1 Microns
  • Low Ionic; low in sodium and potassium
  • Low non-volatile residue levels
  • Biodegradable
  • Free-rinsing
Specifications
Product TypeFloor Cleaner
Product LineNovaClean™
SterilityNon-sterile
AppearanceClear
Composition0.1μm filtered low ionic surfactant
For Use With (Application)General Purpose Cleanroom Filtered Detergent, Free-Rinsing
For Use With (Equipment)Mops and Buckets or Wipes and Sponge
Volume (English)1 gal.
Volume (Metric)3.8 L
Unit SizeCS
Contents & Storage
Store at ambient temperature. Avoid direct sunlight.