M-Zone™ SnapMop™ Flat Head Mop Cover
Actual product may vary
Micronova
M-Zone™ SnapMop™ Flat Head Mop Cover
A range of lightweight, snap-on flat head mop covers with urethane foam interiors, designed for effective cleaning and sanitizing in the cleanroom. Available in a variety of fabric options to cope with the most critical cleaning challenges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialDimensionsColor
MICMZSM6AA-18Microfiber Polyester, Nylon, Urethane Foam, Polyester, Polyester45.7 x 12.7 cm (18 x 5 in.)Blue
MICMZSM56-9Smooth Polyester, Polyester, Polyester22.9 x 12.7 cm (9 x 5 in.)White
MICMZSM68-14Polyester Abrasive Mesh, Polyester, Polyester35.6 x 12.7 cm (14 x 5 in.)White
Catalog number MICMZSM6AA-18
Price (SGD)
865.00
Case
Material:
Microfiber Polyester, Nylon, Urethane Foam, Polyester, Polyester
Dimensions:
45.7 x 12.7 cm (18 x 5 in.)
Color:
Blue
Price (SGD)
865.00
Case
The Micronova M-Zone™ SnapMop™ Flat Head Mop Covers fit onto the 360° swivel SnapMop™ Adapter, which can be locked at 180° for more controlled cleaning. A variety of fabric options provide excellent absorbency, effective cleaning and disinfectant dispersal for specialized cleaning. Available in a variety of lengths to match the size of the surface, from equipment cleaning to a large production room.
  • Snap-on flat head mop covers
  • Polyester urethane foam interiors are fully encased in a choice of fabric
  • Stainless steel snap fasteners
  • Available in a range of fabrics for specialized cleaning:
    • NovaPoly — 100% smooth knit polyester provides excellent disinfectant dispersal
    • PolySorb — 100% textured polyester for removing residues and dispersing disinfectant
    • MegaTex — 100% non-woven polyamide is absorbent with a mildly abrasive surface
    • PolyMesh — 100% polyester with a layer of mesh provides a mild scrubbing surface
    • NovaLite — Absorbent chamois fabric with good friction resistance
  • Available without a foam interior for quick wipe downs and sanitization
  • Available in a variety of lengths to match cleaning surface size
    • The 9-inch covers are excellent for pass-throughs and smaller spaces
    • The 14 and 18-inch covers are ideal for floors, walls and ceilings
  • Fits on QPSL plastic frame or QDSL stainless steel frame
  • Swivel joint rotates 360° but can be locked at 180° for more controlled cleaning
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
Specifications
Dimensions45.7 x 12.7 cm (18 x 5 in.)
Product TypeMop Cover
Head TypeFlat
ColorBlue
Length (English)18 in.
SterilityNon-sterile
Length (Metric)45.7 cm
AutoclavableAutoclavable
Width (Metric)12.7 cm
Width (English)5 in.
Resistant ToChemicals
DisposableReusable
Quantity24/Cs.
Care InstructionsItem may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
IncludesPolyurethane Foam Interior
For Use With (Application)Cleaning and Sanitization of Cleanroom Surfaces
Color-codedNo
MaterialMicrofiber Polyester, Nylon, Urethane Foam, Polyester, Polyester
Unit SizeCase