M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser
M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser
Actual product may vary
M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser
Touch-free, hand care dispensing system that offers hassle-free, wall-mount installation and a gravity-fed pump, for simple bottle replacement and efficient battery use.
Have Questions?
Catalog NumberVolume (Metric)Color
MICICCAP-17IR1 mLBlue
Catalog number MICICCAP-17IR
Price (SGD)
66.00
PK
Add to cart
Volume (Metric):
1 mL
Color:
Blue
Price (SGD)
66.00
PK
Add to cart
Touch-free dispenser is designed for use with a choice of ICCAP Dispensing Capsules that screw onto the corresponding 500 mL replaceable bottles and snap into the well of the dispenser. When the electronic beam is broken by a hand passing under the unit, the bubble on the spout depresses to dispense either 1 or 2 mL of hand care into the palm.
  • Touch-free hand care dispenser
  • Wall mount with drip tray
  • Hassle-free installation and bottle replacement
  • Compact, gravity-fed unit, without the need for tubing
  • Efficient battery use
  • For use with a choice of ICCAP Dispensing Capsules:
    • ICCAP-16 - Green 2 mL dispensing capsule for soaps and sanitizer
    • ICCAP-17IR - Blue 1 mL dispensing capsule for sterile IPA or sanitizer
  • Designed for use with the Micronova hand care 500 mL replacement bottles:
    • Hy-G-Clenz - antibacterial soap, Aquahol - sterile alcohol, NovaClenz - hand sanitizer and LoNa - low sodium hand soap
Specifications
Product TypeDispensing Capsule
Color-codedYes
FormCapsules
Tip TypeSpout
ScentUnscented
SterilitySterile
AppearanceColorless
AutoclavableNot Autoclavable
Care InstructionsOnce the spout is attached the inverted bottle snaps into the well of the dispenser. When the electronic beam is broken by a hand passing under the unit, the bubble on the spout depresses to dispense the soap or lotion into the palm.
Certifications/ComplianceISO 9001 Certified Facility Certificate of Conformance Certificate of Irradiation
ColorBlue
Dimensions (L x W x H)1.375 x 1.375 x 3.25 in. (3.49 x 3.49 x 8.26 cm)
DisposableReusable
FilteredNon-filtered
FlammableNo
For Use With (Application)A bubble spout screws onto the 500 ml bottle to use with sterile IPA
For Use With (Equipment)M-Zone MicroDispenser
Height (English)3.25 in.
Height (Metric)8.26 cm
Length (English)1.375 in.
Length (Metric)3.49 cm
MaterialPolypropylene
Quantity6/Pk.
Resistant ToResistant to most chemicals
Size5 x 5 x 5
Volume (English)0.03 oz.
Volume (Metric)1 mL
Width (English)1.375 in.
Width (Metric)3.49 cm
Unit SizePK