Hand Soaps and Sanitizers
Actual product may vary
Hand Soaps and Sanitizers
Hand Soaps and Sanitizers
Micronova
Hand Soaps and Sanitizers
Soaps and sanitizers formulated with highly effective, non-irritating ingredients are ideal for frequent use in clean and controlled environments. Designed for use with the MicroDispenser™. Include a saddle pump.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeSterility
MICIC-220Gloved Hand and Surface SanitizerSterile
MICIC-210Hand SanitizerNon-sterile
MICIC-110Hand and Glove SoapNon-sterile
Catalog number MICIC-220
Price (SGD)
471.00
Case
Product Type:
Gloved Hand and Surface Sanitizer
Sterility:
Sterile
Price (SGD)
471.00
Case
Soaps and sanitizers formulated with highly effective, non-irritating ingredients are ideal for frequent use in clean and controlled environments. Designed for use with the MicroDispenser™. Include a saddle pump.
Hand care products are gentle and effective with built-in skin emollients for frequently washed hands. The 500 mL saddle pump bottles can be used as stand-alone dispensers or with the M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser (ICDISP-3) to ensure uncontaminated results.
  • NovaClenz™ Instant Hand Sanitizer, made with 62% Ethanol to help prevent viral transfer. Effective against bacteria, including: pseudomonas, salmonella and staphylococcus. Effective in reducing nosocomial infections. Fortified with emollients to moisturize hands.
  • Hy-G-Clenz™ Antimicrobial Hand Soap, made with 0.13% Benzalkonium Chloride and natural emollients. Effective against a wide range of both gram-positive and gram-negative bacteria. Persistent activity up to three hours. pH balanced and contains natural moisturizers, ideal for frequent hand washing. Meets OSHA standards.
  • AquaHol™ Sterile Isopropyl Alcohol Solution, made with 70% Sterile IPA in 30% Wifi Quality Water, and meets USP requirements. Terminally filtered to 0.1 microns. Gamma Irradiated. Ideal for use over cleanroom gloves.
  • NovaDyne™ General Purpose Hand Soap, pH balanced gentle lotion hand soap. Made with built-in moisturizers/skin emollients, ideal for frequent hand washing.
  • LoNa™ Low Sodium Hand Soap, low ionic, low sudsing hand soap and glove cleanser, specially designed for the high-tech cleanroom; less than 100 ppb sodium. pH balanced and clean enough to be used on the face.
Specifications
Product TypeGloved Hand and Surface Sanitizer
Color-codedNo
FormClear Liquid
pH7
ScentRegular Scent
SterilitySterile
AppearanceColorless
AutoclavableNot Autoclavable
Boiling Point83°C (181.4°F)
Care InstructionsIdeal for use over cleanroom gloves prior to entering into aseptic areas and between critical processes.
Certifications/ComplianceOSHA, Analysis, Conformance, Irradiation
ColorClear
Composition70% IPA/30% WFI Quality Water
Concentration70% IPA
Container TypeHDPE
Density0.872 to 0.883 at 20°C
Dimensions (L x W x H)7.75 x 1.75 x 3.75
DisposableSingle-use
FilteredFiltered
FlammableYes
For Use With (Application)Sterilizing agent for hands and gloves
For Use With (Equipment)ICDISP-3 Touch-free MicroDispenser
Height (English)7.75 in.
Height (Metric)19.69 cm
Length (English)3.75 in.
Length (Metric)9.53 cm
Melting Point-90°C (-130°F)
Packaging6 bottles per poly-lined case
Quantity6/Cs.
Resistant ToChemical
Shelf Life36 Months
Size500 mL
SolubilitySoluble in water
Vapor Pressure4.1 kPa (30.753 mmHg) at 20°C
Volume (English)16.9 oz.
Volume (Metric)500 mL
Weight (English)16.9 oz.
Weight (Metric)0.5 kg
Width (English)1.75 in.
Width (Metric)4.45 cm
Unit SizeCase
Contents & Storage
Store away from all heat/ignition sources and in original container that is tightly closed. Store locked up and separate from oxidizing materials. Avoid direct sunlight. Empty containers may retain product residue and can be hazardous.