Hand Soaps and Sanitizers
Actual product may vary
Hand Soaps and Sanitizers
Hand Soaps and Sanitizers
Micronova
Hand Soaps and Sanitizers
Soaps and sanitizers formulated with highly effective, non-irritating ingredients are ideal for frequent use in clean and controlled environments. Designed for use with the MicroDispenser™. Include a saddle pump.
Have Questions?
Catalog NumberProduct TypeSterility
MICIC-210Hand SanitizerNon-sterile
Catalog number MICIC-210
Price (SGD)
262.00
Case
Product Type:
Hand Sanitizer
Sterility:
Non-sterile
Price (SGD)
262.00
Case
Soaps and sanitizers formulated with highly effective, non-irritating ingredients are ideal for frequent use in clean and controlled environments. Designed for use with the MicroDispenser™. Include a saddle pump.
Hand care products are gentle and effective with built-in skin emollients for frequently washed hands. The 500 mL saddle pump bottles can be used as stand-alone dispensers or with the M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser (ICDISP-3) to ensure uncontaminated results.
  • NovaClenz™ Instant Hand Sanitizer, made with 62% Ethanol to help prevent viral transfer. Effective against bacteria, including: pseudomonas, salmonella and staphylococcus. Effective in reducing nosocomial infections. Fortified with emollients to moisturize hands.
  • Hy-G-Clenz™ Antimicrobial Hand Soap, made with 0.13% Benzalkonium Chloride and natural emollients. Effective against a wide range of both gram-positive and gram-negative bacteria. Persistent activity up to three hours. pH balanced and contains natural moisturizers, ideal for frequent hand washing. Meets OSHA standards.
  • AquaHol™ Sterile Isopropyl Alcohol Solution, made with 70% Sterile IPA in 30% Wifi Quality Water, and meets USP requirements. Terminally filtered to 0.1 microns. Gamma Irradiated. Ideal for use over cleanroom gloves.
  • NovaDyne™ General Purpose Hand Soap, pH balanced gentle lotion hand soap. Made with built-in moisturizers/skin emollients, ideal for frequent hand washing.
  • LoNa™ Low Sodium Hand Soap, low ionic, low sudsing hand soap and glove cleanser, specially designed for the high-tech cleanroom; less than 100 ppb sodium. pH balanced and clean enough to be used on the face.
Specifications
Product TypeHand Sanitizer
Color-codedNo
FormGel
pH7
ScentUnscented
SterilityNon-sterile
AppearanceClear
AutoclavableNot Autoclavable
Boiling Point77.78°C (172°F)
Care InstructionsKeep at ambient temprature. Avoid direct sunlight. Keep away from heat, hot surfaces, sparks, open flames and other ignition.
Certifications/ComplianceCertificate of Analysis
ColorClear
Composition60% Ethanol Sanitizing Gel
Concentration65 to 72%
Container TypeHDPE
Density0.89 g/mL at 20°C
Dimensions (L x W x H)7.75 x 1.75 x 3.75
DisposableSingle-use
FilteredNon-filtered
FlammableYes
For Use With (Application)Reduces incidence of nosocomial infections
Height (English)7.75 in.
Height (Metric)19.69 cm
Length (English)3.75 in.
Length (Metric)9.53 cm
PackagingPolyethylene lined carton
Quantity6/Cs.
Shelf Life36 Months
Size500 mL
SolubilityEasily soluble in cold water and hot water.
StyleStackable
Viscosity47,000 and 150,000 mPa.s.
Volume (English)16.9 oz.
Volume (Metric)500 mL
Weight (English)16.9 oz.
Weight (Metric)0.5 kg
Width (English)1.75 in.
Width (Metric)4.45 cm
Unit SizeCase
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials. Keep container tightly closed and sealed until ready for use.