Check Order Status
Flashlight Holder - Clamp
Actual product may vary
Flashlight Holder - Clamp
For use with Inspection Mirror to run the unit behind equipment and fixtures to ensure all surfaces have been properly cleaned
Have Questions?
Catalog Number
For Use With (Equipment)
MICFLASHH-01
QDIST-66P (Inspection Mirror)
Catalog number
MICFLASHH-01
Price (SGD)
64.00
EA
Add to cart
For Use With (Equipment):
QDIST-66P (Inspection Mirror)
Price (SGD)
64.00
EA
Add to cart
Use with the highly polished stainless steel mirror for inspecting hard to reach areas
Features swivel connection allowing it to be attached to any SSU series handle
The optional flashlight attachment is ideal for use in tanks and other low light applications
Specifications
Description
Clamp style flashlight holder with 360° rotation
Dimensions (L x W x H)
3.6 x 2.0 x 1.2 in.
For Use With (Equipment)
QDIST-66P (Inspection Mirror)
Material
Plastic
Product Type
Flashlight Clamp