Stainless Steel Compact Cart and Bucket System
Stainless Steel Compact Cart and Bucket System
Actual product may vary
Micronova
Stainless Steel Compact Cart and Bucket System
A compact and highly maneuverable stainless steel cart system with 360° rotation Prevenz Antimicrobial Wheels and smooth, molded stainless steel buckets. Available in single, double and triple bucket configurations.
Have Questions?
Catalog NumberProduct TypeDimensions
MICC-92Double Bucket Cart21 x 25.5 x 17 in.
Catalog number MICC-92
Price (SGD)
-
Product Type:
Double Bucket Cart
Dimensions:
21 x 25.5 x 17 in.
The Micronova Stainless Steel Compact Cart Systems feature electropolished 304 stainless steel carts, available in single, double and triple bucket configurations. The included stainless steel buckets are molded with no ridges or seams to harbor bacteria.
  • Compact and highly maneuverable cart system
  • Electropolished 304 stainless steel cart and buckets
  • Prevenz Antimicrobial Wheels with 360° rotation
  • Available in single, double and triple bucket configurations
  • Durable bacteria-resistant construction
  • Rectangular buckets accept 18 inch wide mops
  • Compatible with MegaWringer and Slim-T Wringer
  • Autoclavable
Specifications
Product TypeDouble Bucket Cart
Capacity (Metric)30.3 L/Bucket, 60.6 L total
MaterialStainless Steel (304), Electropolished, Polyketone
AutoclavableAutoclavable
Capacity (English)8 gal.
Care InstructionsStore at Ambient Temperature. Rinse to remove disinfectant before Autoclave or Storage
Dimensions21 x 25.5 x 17 in.
For Use With (Application)Cleaning Walls, Floors and Ceilings
For Use With (Equipment)Slim-T Wringer series, fits 18 in. W mops
Height (English)17 in.
Height (Metric)43.2 cm
IncludesTwo 8 gal. Buckets
Length (English)21 in.
Length (Metric)53.3 cm
SterilityNon-sterile
Width (English)25.5 in.
Width (Metric)64.8 cm
Unit SizeEA