Irradiated BucketLiners™
Irradiated BucketLiners™
Actual product may vary
Irradiated BucketLiners™
Creates an impermeable barrier to strong solutions and raw materials when used to line cleanroom buckets and storage containers.
Have Questions?
Catalog NumberDimensionsPackaging Type
MICBKL-2430IR60 x 90 cm (24 x 30 in.)Individually Double bagged
Catalog number MICBKL-2430IR
Price (SGD)
450.00
CS
Add to cart
Dimensions:
60 x 90 cm (24 x 30 in.)
Packaging Type:
Individually Double bagged
Price (SGD)
450.00
CS
Add to cart
  • The unique LDPE/ Nylon laminate is a superb barrier material for lining totes to transport raw materials or to line buckets containing disinfectants or other cleaning solutions
  • Flat packed
  • Hold up to autoclave with liquid contents
  • Specifications
    Product TypeCleanroom Sterile Bag
    Capacity (Metric)22 L
    MaterialLDPE
    AutoclavableNot Autoclavable
    Capacity (English)6 gal.
    ColorClear
    DescriptionLaminate: 75% LDPE 25% Nylon
    Dimensions60 x 90 cm (24 x 30 in.)
    DisposableSingle-use
    For Use With (Application)Product Transport or Liquid Containment
    For Use With (Equipment)8-gallon container or Bucket
    Height (Metric)75 cm
    Length (English)24 in.
    Length (Metric)60 cm
    Packaging TypeIndividually Double bagged
    Quantity24/Pk.
    SterilitySterile
    Sterilization MethodGamma-Irradiation
    Thickness (Metric)0.080 mm
    Width (English)30 in.
    Width (Metric)90 cm
    Unit SizeCS