Slimline cleanroom bucket
Slimline cleanroom bucket
Actual product may vary
Slimline cleanroom bucket
Micronova Slimline cleanroom bucket
Have Questions?
Catalog number MICB-7DBAC
Price (SGD)
373.00
EA
Add to cart
Price (SGD)
373.00
EA
Add to cart
A narrow polypropylene single replacement bucket in blue for use with the SlimLine or SlimT Cleanroom Cart system. Each bucket measures 19.25 x 9.5 x 9.5 in. and has a 6 gallon capacity, supplied with casters. Can be autoclaved.
A narrow polypropylene single replacement bucket in blue for use with the SlimLine or SlimT Cleanroom Cart system. Each bucket measures 19 1/4 in. x 9 1/2 in. x 9 1/2 in. and has a 6 gallon capacity. Can be autoclaved.
Specifications
Product TypeBucket
AutoclavableAutoclavable
Includes3 in. Casters
Unit SizeEA