SlimLine™ Buckets
Actual product may vary
Micronova
SlimLine™ Buckets
These rectangular, 6 gallon buckets with a 20-inch width are an ideal choice for use with flat head mops and string mops in controlled environments. Designed for stand alone use or as replacements for Micronova SlimLine™ and SlimT™ Series Carts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
MICB-7DBBlue
MICB-7YEYellow
Catalog number MICB-7DB
Price (SGD)
169.00
Each
Color:
Blue
Price (SGD)
169.00
Each
Micronova SlimLine™ Polypropylene Buckets are designed to work well with Micronova BucketLiners™ and the range of W-9 Series Wringers. The color-coded buckets are available in a choice of red, yellow, blue, green and grey.
  • Rectangular polypropylene buckets
  • 20-inch width is ideal for flat mops and string mops
  • 6 gallon capacity
  • Volume graduations with gallon and liter markers
  • Color-coded with a choice of red, yellow, blue, green and grey
  • Work with BucketLiners and W-9 Series Wringers
  • Replacement buckets for SlimLine and SlimT Series Carts
  • Autoclavable
Specifications
Product TypeMop Bucket
Capacity (Metric)22.7 L
MaterialPolypropylene
AutoclavableAutoclavable
Capacity (English)6 gal.
Care InstructionsStore at Ambient Temperature. To Autoclave - do not stack - invert Bucket to allow drainage.
ColorBlue
Dimensions55.9 x 30.5 x 26.7 cm (22 x 12 x 10.5 in.)
For Use With (Application)Cleaning Walls, Floors and Ceilings
For Use With (Equipment)SlimLine mop and Bucket System
Height (English)10.5 in.
Height (Metric)26.7 cm
Length (English)22 in.
Length (Metric)55.9 cm
SterilityNon-sterile
Width (English)12 in.
Width (Metric)30.5 cm
Unit SizeEach