SlimLine™ Irradiated Flat Head Mop Covers
SlimLine™ Irradiated Flat Head Mop Covers
Actual product may vary
SlimLine™ Irradiated Flat Head Mop Covers
A series of irradiated, elasticized, 100% smooth polyester slipcovers with urethane foam interiors that are ideal for fine cleaning and sanitizing all surfaces and equipment in the cleanroom and controlled environments, ISO 5 to 9.
Have Questions?
Catalog NumberMaterialDimensionsQuantityIncludes
MIC191301235Smooth Knit Polyester, Urethane Foam22.9 x 12.7 cm (9 x 5 in.)12/Cs.Standard Label
Catalog number MIC191301235
Price (SGD)
-
Material:
Smooth Knit Polyester, Urethane Foam
Dimensions:
22.9 x 12.7 cm (9 x 5 in.)
Quantity:
12/Cs.
Includes:
Standard Label
Micronova™ SlimLine™ NovaPoly Irradiated Mop Cover is a polyester/foam slipcover with an elasticized band designed for clean and controlled environments.
The Micronova Irradiated Slimline™ Flat Head Mop Covers fit onto the 360° swivel SlimLine™ Adapters, which can be locked at 180° for more controlled cleaning. These elasticized mop covers easily and securely attach to either the plastic or stainless steel mop frame. Covers are gamma irradiated for aseptic environments.
  • Irradiated, elasticized cleanroom mop covers with urethane foam interiors
  • Easy to change and fits securely on mop frame
  • Ideal for fine cleaning and sanitizing cleanroom surfaces
  • Thorough saturation for ample disinfectant dispersal
  • Foam layer fully encased in polyester
  • NovaPoly — 100% smooth knit polyester provides excellent disinfectant dispersal
  • SMC6 -- Single ply option without foam interior
  • Fits on QPSL, plastic frame or QDSL stainless steel frame
  • Swivel joint rotates 360° but can be locked at 180° for more controlled cleaning
  • Suitable for use with strong disinfectants and repeated autoclaving
  • Gamma irradiated for aseptic environments
  • Launderable and reusable
  • Autoclavable
Specifications
Dimensions22.9 x 12.7 cm (9 x 5 in.)
Product TypeIrradiated Flat Head Mop Cover
ColorWhite
Length (English)9 in.
SterilitySterile
Length (Metric)22.9 cm
AutoclavableAutoclavable
Width (Metric)12.7 cm
Width (English)5 in.
Resistant ToChemicals
DisposableReusable
Quantity12/Cs.
Care InstructionsItem may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
IncludesStandard Label
For Use With (Equipment)SlimLine™ Cleanroom Mop
For Use With (Application)Cleaning and Sanitization of Cleanroom Surfaces
Certifications/ComplianceISO 9001:2008 Facility, 10-6 Sterility Assurance Level, In-compliance with AAMI Guidelines for Sterilization
MaterialSmooth Knit Polyester, Urethane Foam
Unit SizeCS