Product Image
LabServ 12" PF Nitrile Glove, XS, 100/Bx
Have Questions?
Catalog number LVG671260XS
Price (SGD)
20.00
Special offer
Ends: 06-Sep-2026
21.00
Save 1.00 (5%)
Box
Price (SGD)
20.00
Special offer
Ends: 06-Sep-2026
21.00
Save 1.00 (5%)
Box
Specifications
Unit SizeBox