Product Image
LabServ 12" PF Nitrile Glove, XL, 100/Bx
Have Questions?
Catalog number LVG671260XL
Price (SGD)
20.00
Special offer
Ends: 31-Dec-2026
21.00
Save(5%)
Box
Price (SGD)
20.00
Special offer
Ends: 31-Dec-2026
21.00
Save(5%)
Box
Specifications
Unit SizeBox