Sterile 70% IPA 30% WFI with Spray, 1 L
Sterile 70% IPA 30% WFI with Spray, 1 L
Actual product may vary
LabServ
Sterile 70% IPA 30% WFI with Spray, 1 L
70% IPA is a general-purpose disinfectant for use in controlled environments. It is a blend containing 70% USP grade Isopropyl Alcohol (IPA) and 30% USP Water for Injection (WFI).
Have Questions?
Catalog NumberForm
LVA7031Liquid
Catalog number LVA7031
Price (SGD)
200.00
Special offer
Ends: 31-Mar-2026
262.00
Save 62.00 (24%)
Case
Add to cart
Form:
Liquid
Price (SGD)
200.00
Special offer
Ends: 31-Mar-2026
262.00
Save 62.00 (24%)
Case
Add to cart
  • Bag-in-bottle trigger spray is an airtight closed system which maintains sterility and integrity of the content throughout use
  • The integrated collapsible bag eliminates the risk of airborne contamination and possibility of suck back during the use of conventional trigger spray
  • Cost effective - the system allows all content to be dispensed from the bottle to avoid any wastage
  • Double-bagged and double sealed polybag case liner
  • Guaranteed to have an endotoxin level of less than 0.25 EU/mL
  • Individually lot coded for ease of traceability and quality control
Specifications
Color-codedNo
FormLiquid
ScentUnscented
SterilitySterile
Quantity1 L
Unit SizeCase