▸
Flasks
▸
Cell Culture Flasks
▸
Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Actual product may vary
Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Perfect for cell growth, cell yields aim on little or medium input volume.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Culture Area
Closure Type
LSVFB012937
250 mL
75 cm²
Vented
LSVFB012935
50 mL
25 cm²
Vented
FISFB012937
250 mL
75 cm²
Vented
LSVFB012939
600 mL
182 cm
2
Vented
4 Options
Catalog number
LSVFB012937
Price (SGD)
122.00
Special offer
Ends:
31-Dec-2026
168.00
Save 46.00 (27%)
Case
Add to cart
Capacity (Metric):
250 mL
Culture Area:
75 cm²
Closure Type:
Vented
Price (SGD)
122.00
Special offer
Ends:
31-Dec-2026
168.00
Save 46.00 (27%)
Case
Add to cart
Flat, free flask surface
Striation free to maximize usable growth area
Used in both open, closed systems
Angled neck design offers good pipet and cell scraper access
Upper triangular, wider base shape provides stability
Protruding ridge on back side of flask makes it easy for stacking
Special area near neck for easy mark writing
Both flask sides have engraved graduation
Sterilized by gamma irradiation
Non-pyrogenic
Vented closures
Vented cap filter is PVDF; pore size: 0.22μm
Specifications
Certifications/Compliance
CE
Closure Color
Blue
Color
Natural
Capacity (Metric)
250 mL
For Use With (Application)
Cell growth
Neck Style
Angled
No. per Case
100 Flasks
No. per Pack
5 Flasks
Product Type
Tissue Culture Flask
Surface Coating
Surface-treated
Writing Surface
Flat
Graduated
Yes
Sterility
Sterile
Bottom Shape
Flat
Closure Type
Vented
Culture Area
75 cm²