Surface Treated SterileTissue Culture Plates
Surface Treated SterileTissue Culture Plates
Actual product may vary
Surface Treated SterileTissue Culture Plates
Surface Treated SterileTissue Culture Plates
Surface Treated SterileTissue Culture Plates
Surface Treated SterileTissue Culture Plates
Ideal for comparison and analysis of cell growth, yield in multiple platforms.
Have Questions?
Catalog NumberCulture Area
LSVFB0129300.83 cm2
Catalog number LSVFB012930
Price (SGD)
114.21
Special offer
Ends: 15-Dec-2025
178.00
Save 63.79 (36%)
CS
Add to cart
Culture Area:
0.83 cm2
Price (SGD)
114.21
Special offer
Ends: 15-Dec-2025
178.00
Save 63.79 (36%)
CS
Add to cart
  • Uniform well volume to ensure equal growth surface area
  • Well surface smooth, free from striation to maximize usable growth area
  • Raised rims on wells with uniform rings on lid to reduce evaporation
  • Single position lid reduces risks of cross-contamination, handling mistakes
  • Alphanumeric code on wells for easy identification
  • Suitable for all common instruments, automation
  • Sterilized by gamma irradiation
  • Non-pyrogenic
Specifications
Array6 x 8
Certifications/ComplianceCE
Bottom ShapeFlat
Capacity (Metric)1.55 mL
ColorNatural
Culture Area0.83 cm2
MaterialVirgin Polystyrene
ShapeRaised Rims with Condensation Rings
SterilitySterile
Surface TreatmentStandard
TypeTissue Culture Plate
Well Count48
For Use With (Application)Tissue culture
No. per Pack100 Plates
PackagingIndividually wrapped
Unit SizeCS