Product Image
Serological Pipet 50ml, Sterilized, Non-
Have Questions?
Catalog number LSV14955158
Price (SGD)
61.00
Special offer
Ends: 06-Sep-2026
88.00
Save 27.00 (31%)
Case
Price (SGD)
61.00
Special offer
Ends: 06-Sep-2026
88.00
Save 27.00 (31%)