Product Image
Serological Pipet 25ml, Sterilized, Non-
Have Questions?
Catalog number LSV14955157
Price (SGD)
56.00
Special offer
Ends: 06-Sep-2026
81.00
Save 25.00 (31%)
Case
Price (SGD)
56.00
Special offer
Ends: 06-Sep-2026
81.00
Save 25.00 (31%)