Type I Fiberglass Step Stool
Actual product may vary
LabServ
Type I Fiberglass Step Stool
Duty rating of 250 lb. and features a molded top that provides a large standing platform.
Have Questions?
Catalog NumberLength (English)
LSG600224 in.
Catalog number LSG6002
Price (SGD)
-
Length (English):
24 in.
HolsterTop™ with Lock-In System secures tools to increase productivity
  • Slip-resistant Traction-Tred™ Steps
  • Braces on bottom steps
  • The blue and yellow ladder is an exclusive registered trademark of Werner Co
Specifications
Certifications/ComplianceANSI-A14-2-2007, OSHA
Dimensions (L x W x H)24 x 17.28 x 6.24 in.
Height (English)6 ft.
Length (English)24 in.
Model6002 Series
Quantity1 Ea.
Width (English)17.2 in.
Load Capacity (English)250 lb.
Load Capacity (Metric)113.4 kg
MaterialFiberglass
Product TypeStep Stool
Unit SizePack