Barnstead™ Easypure™ RoDi Accessories and Consumables
Barnstead™ Easypure™ RoDi Accessories and Consumables
Actual product may vary
Barnstead™ Easypure™ RoDi Accessories and Consumables
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
LMX13
also known as BARNSTEAD = LMX13
Replacement Ultraviolet Lamp
D502133Cartridge Kit
Catalog number LMX13
also known as BARNSTEAD = LMX13
Price (SGD)
458.00
EA
Add to cart
Description:
Replacement Ultraviolet Lamp
Price (SGD)
458.00
EA
Add to cart
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
Specifications
DescriptionReplacement Ultraviolet Lamp
For Use With (Equipment)Barnstead Easypure RoDi
TypeUV Lamp
Unit SizeEA