Sparkfree Freezer, 124L
Sparkfree Freezer, 124L
Actual product may vary
Sparkfree Freezer, 124L
The Labcold Sparkfree range has a specially manufactured sparkfree interior with all sources of potential ignition removed from the chamber. Anything that could cause a spark is located outside of the chamber making it an important part of laboratory safety.
Have Questions?
Catalog NumberCapacity (Metric)
LCDRLVF0417124 L
Catalog number LCDRLVF0417
Price (SGD)
-
Capacity (Metric):
124 L
  • Sparkfree safety interior
  • Manual defrost
  • Door Lock
  • Access Port
  • Self closing door
  • Temperature display
  • High/Low temperature alarm
  • Door Open alarm
  • 3 shelves
Specifications
TypeUnderbench
Capacity (Metric)124 L
Temperature Range-18°C to -25°C
Door StyleSolid
Shelves3
Dimensions (L x W x H) Exterior 600 x 600 x 830 mm
Unit SizeEach