150L Sparkfree Refrigerator
150L Sparkfree Refrigerator
Actual product may vary
150L Sparkfree Refrigerator
150L Sparkfree Refrigerator
150L Sparkfree Refrigerator
The Labcold™ Sparkfree Refrigerator has a specially manufactured sparkfree interior with all sources of potential ignition removed from the chamber. Anything that could case a spark is located outside of the chamber making it a significant aid to laboratory safety.
Have Questions?
Catalog NumberCapacity (Metric)
LCDRLPR0517150 L
Catalog number LCDRLPR0517
Price (SGD)
-
Capacity (Metric):
150 L
  • Sparkfree safety interior
  • Off Cycle Automatic defrosting
  • 3 adjustable shelves
  • Door lock
  • Temperature alarm and display
  • High/Low temperature alarm
  • Door Open alarm
  • Access port
  • Touch screen controller
Specifications
Capacity (Metric)150 L
Temperature Range0°C to 10°C
Door StyleSolid
No. of Doors1
Shelves3 Adjustable
StyleUndercounter
PortsYes
DefrostAutomatic
Depth (Metric) Exterior600 mm
Height (Metric) Exterior835 mm
Width (Metric) Exterior600 mm
Weight (Metric)45 kg
Unit SizeEach