Biological Safety Cabinet/Laboratory Cart Accessory, Logic Cart
Biological Safety Cabinet/Laboratory Cart Accessory, Logic Cart
Actual product may vary
Biological Safety Cabinet/Laboratory Cart Accessory, Logic Cart
Labconco Logic Cart is specifically designed to provide convenient storage of supplies commonly used in biological safety cabinets.
Have Questions?
Catalog number LBC8022000
Price (SGD)
-
Labconco Logic Cart is specifically designed to provide convenient storage of supplies commonly used in biological safety cabinets.
  • Fits under Logic Series enclosures
  • 2 in. casters
  • Work surface: polyethylene, 19L x 25 in.W
  • Frame: ABS
  • Cart comprises drawer, three side compartments and open storage area
Specifications
Product TypeLogic Cart
ColorWhite
Depth (English)19 to 21.5 in.
Depth (Metric)44.2 to 48 cm
Dimensions (L x W x H)25 x 19 x 27 in. (63.5 x 48.3 x 68.6 cm)
For Use With (Equipment)PuriCare procedure stations, purifier logic biosafety cabinets
Height (English)27 in.
Height (Metric)68.6 cm
IncludesDrawer, 3 side compartments and open storage area
Length (English)25 in.
Length (Metric)63.5 cm
MaterialPolyethylene
No. of Drawers2
No. of Shelves5
Weight (Metric)22.7 kg
Weight (English)150 lb.
Width (English)19 in.
Width (Metric)48.3 cm
Unit SizeEA