Secondary Trap Canister and Inserts for CentriVap™ System
Secondary Trap Canister and Inserts for CentriVap™ System
Actual product may vary
Secondary Trap Canister and Inserts for CentriVap™ System
Help to provide secondary protection to vacuum pumps in evaporation systems
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeWeight (English)Weight (Metric)
LBC7814800Acid Trap Insert4 lb.1.8 kg
LBC7460900Secondary Trap Canister5 lb.2.3 kg
Catalog number LBC7814800
Price (SGD)
353.00
EA
Add to cart
Type:
Acid Trap Insert
Weight (English):
4 lb.
Weight (Metric):
1.8 kg
Price (SGD)
353.00
EA
Add to cart
Help to provide secondary protection to vacuum pumps in evaporation systems
  • Clear polypropylene canister with mounting bracket, 17 in. (43cm) PVC tubing, and two clamps
  • Canister accommodates trap inserts listed in the table
Specifications
MaterialPolypropylene
For Use With (Equipment)CentriVap™ system
TypeAcid Trap Insert
Weight (English)4 lb.
Weight (Metric)1.8 kg
Unit SizeEA