Drying Chamber for FreeZone™ Freeze-Dry Systems, PTFE-coated
Drying Chamber for FreeZone™ Freeze-Dry Systems, PTFE-coated
Actual product may vary
Drying Chamber for FreeZone™ Freeze-Dry Systems, PTFE-coated
Drying Chamber with corrosion-resistant interior for use on FreeZone™ Freeze Dryers.
Have Questions?
Catalog NumberNo. of Ports
LBC752280112
Catalog number LBC7522801
Price (SGD)
7,031.00
EA
Add to cart
No. of Ports:
12
Price (SGD)
7,031.00
EA
Add to cart
  • Drying chamber made of type 304 stainless steel with corrosion-resistant PTFE-coated interior
  • Includes acrylic lid with neoprene gasket, latching pin, and neoprene valves with molded plastic knobs for 1/2 in. and 3/4 in. adapters
  • For use with FreeZone™ 1, 2.5, 4.5, 6, 8, 12, or 18 Liter Freeze Dryers
Specifications
TypeDrying Chamber
No. of Shelves0
MaterialPTFE-coated Stainless Steel
No. of Ports12
For Use With (Application)To provide a means to attach flasks to freeze dry system or contain samples in bulk
For Use With (Equipment)FreeZone™ Freeze Dryers
Diameter (English)8 in.
Height (English)9.75 in.
Weight (English)10 lb.
Unit SizeEA