Centrivap Ultra Low Cold Trap
Centrivap Ultra Low Cold Trap
Actual product may vary
Centrivap Ultra Low Cold Trap
The CentriVap -84°C Cold Trap collects evaporated liquid from a CentriVap Concentrator, protecting an accessory vacuum pump and the laboratory environment from the corrosive effects of vapors and fumes emitted from samples.
Have Questions?
Catalog number LBC7460035
Price (SGD)
18,569.00
EA
Add to cart
Price (SGD)
18,569.00
EA
Add to cart
The CentriVap -84°C Cold Trap collects evaporated liquid from a CentriVap Concentrator, protecting an accessory vacuum pump and the laboratory environment from the corrosive effects of vapors and fumes emitted from samples.
  • Brushed, Stainless Steel and Powder-Coated Steel Exterior
  • Illuminated Temperature Graph with Endpoint Indication (Operating temperature of -40°C)
  • HCFC-Free Refrigerant System
  • 1/3 hp Compressor (Reaches -84°C in less than 40 min.)
  • 4-liter, Stainless Steel Collector
  • Acrylic or Stainless Steel Lid (with 2 Quick-Disconnect fittings for connection to a CentriVap Concentrator)
  • Stainless Steel Drain Port (with corrosion-resistant, nylon valve)
  • Recommended for Solvent (e.g. Acetone, Chloroform) sample applications
    Specifications
    Certifications/ComplianceCE Marked
    MaterialStainless Steel
    For Use With (Equipment)Low Freezing Point Solvents
    Plug TypeBritish (UK)
    Temperature (Metric) Operating-40°C
    Volume (Metric)4 L
    Dimensions (L x W x H)14.5 x 22.0 x 20.9 in. (36.8 x 55.9 x 53.1 cm)
    Length (English)14.5 in.
    Length (Metric)36.8 cm
    Width (English)22.0 in.
    Width (Metric)55.9 cm
    Height (English)20.9 in.
    Height (Metric)53.1 cm
    Electrical Requirements230 V 50 Hz 6 amp (International)
    Voltage0 to 300 V
    Unit SizeEA