Tube Holders for Fast-Freeze™ and Lyph-Lock™ Flasks
Tube Holders for Fast-Freeze™ and Lyph-Lock™ Flasks
Actual product may vary
Tube Holders for Fast-Freeze™ and Lyph-Lock™ Flasks
Holds tubes in upright position for freeze drying.
Have Questions?
Catalog NumberHeight (English)Height (Metric)
LBC74427005 in.13.9 cm
Catalog number LBC7442700
Price (SGD)
731.00
EA
Add to cart
Height (English):
5 in.
Height (Metric):
13.9 cm
Price (SGD)
731.00
EA
Add to cart
Holds tubes in upright position for freeze drying.
Hold tubes in upright position inside 900mL or larger Fast-Freeze Flasks and 750mL or larger Lyph-Lock Flasks Stainless steel
Specifications
TypeTube Holder
Capacity (Metric)750, 900 mL
MaterialStainless Steel
For Use With (Application)Hold tubes in upright position
For Use With (Equipment)900ml or larger fast freeze™ flasks and 750ml or larger lyph lock flasks
Diameter (English)3.5 in.
Diameter (Metric)8.33 cm
Height (English)5 in.
Height (Metric)13.9 cm
Weight (English)0.5 lb.
Weight (Metric)0.5 kg
Unit SizeEA