Lids for FreeZone™ Freeze Dryers
Lids for FreeZone™ Freeze Dryers
Actual product may vary
Lids for FreeZone™ Freeze Dryers
Replacement lids for FreeZone™ Freeze Dryer collector chambers.
Have Questions?
Catalog NumberFor Use With (Equipment)Material
LBC7207500FreeZone™ 2.5L Freeze DryersStainless Steel
Catalog number LBC7207500
Price (SGD)
643.00
EA
Add to cart
For Use With (Equipment):
FreeZone™ 2.5L Freeze Dryers
Material:
Stainless Steel
Price (SGD)
643.00
EA
Add to cart
Stainless Steel, 14 in.
  • Stainless steel lid has 14.0 in. diameter with 3 in. diameter hole
  • Optional collector lid for FreeZone™ 4.5L, 6L, 8L, 12L and 18L Freeze Dryers
  • For use with solvents that attack arcylic, like acetonitrile
  • Includes neoprene gasket
Specifications
TypeChamber Lid
MaterialStainless Steel
For Use With (Equipment)FreeZone™ 2.5L Freeze Dryers
IncludesNeoprene gasket
Diameter (English)10.12 in.
Height (English)1.25 in.
Height (Metric)3.18 cm
Length (English)10.38 in.
Length (Metric)26.37 cm
Shipping Weight (English)5 lb.
Weight (English)3 lb.
Unit SizeEA