Kimtech™ G5 Sterile Latex Hand Specific Gloves
Product Image
Kimtech™ G5 Sterile Latex Hand Specific Gloves
Kimtech™ G5 Sterile Latex gloves provide sensitive and dextrous protection in EU GMP ISO Class 5 Grade A sterile cleanrooms. The hand-specific gloves are suitable for double-donning and feature a textured finish for enhanced tactile performance.
Have Questions?
Catalog number KLCHC1170S
Price (SGD)
-
Kimtech™ G5 Sterile Latex gloves provide sensitive and dextrous protection in EU GMP ISO Class 5 Grade A sterile cleanrooms. The hand-specific gloves are suitable for double-donning and feature a textured finish for enhanced tactile performance.
Specifications
ColorNatural
MaterialLatex
Size7