A40 Liquid and Particle Protection Coveralls
Actual product may vary
Ansell
A40 Liquid and Particle Protection Coveralls
Improves safety and comfort in high-risk areas providing general protection against water-based or chemical splashes. Kimberly-Clark™ KleenGuard™ A40 Liquid and Particle Protection Coveralls have the added benefit of anti-static protection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC97920Large
KLC97930X-Large
Catalog number KLC97920
Price (SGD)
-
Size:
Large
  • White in color
  • Single use, disposable
  • Silicone-free and anti-static
  • Specially designed hood to fit a respirator
  • Helps to reduce contamination in low lint and critical areas
  • Full-length zip, over-locked stitched seams and double-layer fabric
  • Ideal for protection against larger particles, radioactive dust and chemical splashes
  • Suited for pharmaceutical areas, manufacturing, utilities, electronics, agriculture and paint-spraying areas
Specifications
Product TypeGeneral Purpose Coveralls
SizeLarge
GenderUnisex
MaterialSpunbond Polypropylene
ColorWhite
DisposableSingle-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Hood TypeElastic
Closure TypeFront Zipper
Seam TypeStitched
ResistanceWater, Chemicals, Liquid, Particle
Latex FreeYes
Length (English)15.6 in.
Length (Metric)40 cm
Weight (English)15.73 lb.
Weight (Metric)7.15 kg
Quantity25/Pk.
Unit SizeCase