KleenGuard™ A40 Coveralls
Actual product may vary
KleenGuard™ A40 Coveralls
Designed to provide protection against larger particles, radioactive dust, and chemical splashes. Use these coveralls in low lint, and critical areas to help reduce contamination.
Have Questions?
Catalog number KLC97910
Price (SGD)
-
Coveralls are designed to provide protection against larger particles, radioactive dust, and chemical splashes. Use these coveralls in low lint, and critical areas to help reduce contamination. Features include a full length zipper, strong over-locked stitched seams, and a hood that is specially designed to fit a respirator.
  • Coveralls offer ultra-low-linting, anti-static (EN 1149-1) and silicon-free protection
  • Film laminate technology provides protection against a wide range of water-based or chemical splashes, liquids, and dust particles
  • Stored in sealed poly bags to reduce the risk of contamination
  • Approved for radioactive dust protection per EN 1073-2
  • This product does not provide protection against radioactive radiation
Specifications
Product TypeSingle-use Coveralls
SizeMedium
GenderUnisex
MaterialSMS, SFL
ColorWhite
DisposableSingle-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Hood TypeElastic
Closure TypeZipper
Seam TypeStitched
SterilityNon-sterile
Protection ClassType 5, Type 6
ResistanceParticle
No. of PocketsNone
StyleFull length zipper
Unit SizeCase