A7 P+™ Laboratory Coats
A7 P+™ Laboratory Coats
Actual product may vary
A7 P+™ Laboratory Coats
A7 P+™ Laboratory Coats
A7 P+™ Laboratory Coats
A7 P+™ Laboratory Coats
A7 P+™ Laboratory Coats
Ansell
A7 P+™ Laboratory Coats
With a cloth-like feel and a reduced risk of heat stress, these lab coats ensure contamination control during extensive periods of use. Kimtech™ A7 P+™ Laboratory Coats made from non-woven SMS fabric with triple stitched seams and provided silicon- and latex-free.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC97710Medium
KLC97720Large
KLC97700Small
KLC97730X-Large
Catalog number KLC97710
Price (SGD)
126.00
Special offer
Ends: 31-Mar-2026
165.00
Save 39.00 (24%)
CS
Add to cart
Size:
Medium
Price (SGD)
126.00
Special offer
Ends: 31-Mar-2026
165.00
Save 39.00 (24%)
CS
Add to cart
Assured Compliance:
  • PPE Cat III according to (EU) Regulation 2016/425 :Type 6 (PB) limited chemical splash protection
  • EN1149-1 Antistatic clothing
  • Tensile strength EN ISO 13934-1 Class 1 of 3
  • Surface resistivity EN 1149-1 < 5 x 1010 ohm
  • Particle shedding test (Helmke Drum) IEST-RP-CC003.3 Category II
  • Bacterial Filtration Efficiency (3.0 μm) ASTM F2100 >99%
Features:
  • High quality disposable laboratory coats with triple stitched seams and breathable, multi-layered material.
  • Enhanced all-round protective features are included, such as high mandarin collars, thumb-loops and extra-length arms to fit lab gloves over coats.
  • The breathable coat material includes high levels of particle and bacterial protection (with filtration efficiency of 99%).
  • The coats are individually packed making them suitable for ISO 7 (Grade C) areas.
  • Anti-slip and low lint material.
  • Type 6 (PB) limited chemical splash protection
Specifications
Product TypeChemical Resistant Lab Coat
SizeMedium
MaterialSMS, Polypropylene, Polyethylene Laminate
ColorWhite
Closure TypeZipper
Cuff StyleElastic
DisposableReusable
SterilityNon-sterile
ResistanceNone
StyleBelow Hip-length
No. of PocketsNone
Certifications/ComplianceCertified PPE Cat III directive 89/686/EEC, EN1149-1:1995 Antistatic clothing, Double Bagged for ISO 7 Grade C areas
Unit SizeCS