1x1 image pixel for data collection
Kimtech Pure™ A5 Non-Sterile Cleanroom Boot Covers
Kimtech Pure™ A5 Non-Sterile Cleanroom Boot Covers
Actual product may vary
Kimtech Pure™ A5 Non-Sterile Cleanroom Boot Covers
Made of 100% polypropylene breathable SMS fabric.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC88850Universal
FIS19-812-864Universal
Catalog number KLC88850
Price (SGD)
-
Size:
Universal
  • Suitable for ISO Class 5 or higher cleanrooms
  • Cloth-like feel
  • Tunneled elastic and bound seams
  • Triple bagged
  • Latex-free
  • Clean processed with Clean-Don™ Technology
  • Vinyl soles
  • Ties for secure fit
Specifications
SizeUniversal
MaterialSMS
Capacity (Metric)5 mL
Latex FreeYes
ConcentrationSlurry: 10mg/mL, 1% solids
ColorWhite
DisposableSingle-use
SterilityNon-sterile
Closure TypeTies
Packaging Quantity100 Pairs/Cs.
Packaging TypeTriple Bagged, Vacuum Packed
Product TypeControlled Environments Boot Covers
Cleanroom ClassClass 100 (ISO 5)
Particle Size1 μm
FormLiquid
Product LinePierce™
Quantity5 mL
Protein FormRecombinant
TargetAntibody
TypeMagnetic Affinity Bead
Ligand TypeProtein L
Unit SizeCase
Contents & Storage
Upon receipt store at 4°C. Product shipped with an ice pack.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link