Kimtech Pure™ A5 Sterile Cleanroom Apparel
Actual product may vary
Ansell
Kimtech Pure™ A5 Sterile Cleanroom Apparel
100% polypropylene breathable SMS fabric has cloth-like feel
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC88803X-Large
FIS19-057-475Small
KLC88800Small
FIS19-057-476Medium
FIS88-801Medium
KLC88801Medium
FIS19-057-490Large
KLC88802Large
FIS19-057-491X-Large
FIS19-057-4922X-Large
Catalog number KLC88803
Price (SGD)
510.00
Special offer
Ends: 06-Sep-2026
618.00
Save 108.00 (17%)
Case
Size:
X-Large
Price (SGD)
510.00
Special offer
Ends: 06-Sep-2026
618.00
Save 108.00 (17%)
Case
100% polypropylene breathable SMS fabric has cloth-like feel
Kimtech™ A5 Sterile Cleanroom Coveralls have an average bacterial filtration efficiency (BFE) of 96% and are made of 100% polypropylene breathable SMS fabric, which gives the wearer cloth-like comfort.
The Reflex Coverall Design is 7.5 times more rip-resistant than ANSI minimum standards and offers excellent fit and freedom of movement for the wearer. This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms.
It is not made with natural rubber latex, silicone, or the BHT preservative and is designed for single-use (disposable PPE). The easy-to-don, individually packaged coveralls feature a mandarin collar, thumb loops and tunneled elastics.
  • 25 Kimtech™ A5 Sterile Cleanroom Coveralls/Case
  • Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms; CE certified (Category III)
  • Reflex Coverall Design offers excellent fit and freedom of movement and 7.5 times more rip-resistant than ANSI minimums
  • This protective apparel isn't made with natural rubber latex, BHT preservative or silicone
  • Kimtech™ A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
Specifications
SizeX-Large
MaterialPolypropylene, SMS
ColorWhite
Cuff StyleElastic
Closure TypeSnap
Seam TypeBound
SterilitySterile
DescriptionTunnelized elastic, built-in thumb loop, folded inside out pattern or cuffed
For Use With (Application)ISO Class 5 or higher cleanroom environment
Unit SizeCase