Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Face Masks
▸
Kimtech™ M3 Sterile Face Masks
Actual product may vary
Kimtech™ M3 Sterile Face Masks
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
Have Questions?
Change view
Catalog Number
Fastening Type
Includes
KLC62470
Earloops
Knitted Earloops
FIS18-999-2405
Ties
7 in. Soft Ties
KLC62467
Ties
7 in. Soft Ties
KLC62494
Ties
9 in. Soft Ties
4 Options
Catalog number
KLC62470
Price (SGD)
200.00
Special offer
Ends:
31-Dec-2026
247.00
Save 47.00 (19%)
Case
Add to cart
Fastening Type:
Earloops
Includes:
Knitted Earloops
Price (SGD)
200.00
Special offer
Ends:
31-Dec-2026
247.00
Save 47.00 (19%)
Case
Add to cart
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
Gamma-sterilized to 10
-6
White with soft ties or earloops
Outer layer of apertured polyethylene film to provide protection from contamination
Polyester binding and polyester filter media provide bacteria filtration efficiency of 91.9% and particle filtration efficiency of 93.1%
Specifications
Product Type
Cleanroom Face Mask
Color
White
Fastening Type
Earloops
Material
SMS
Size
Standard
Sterility
Sterile
Style
Pleated
Cleanroom Class
Class 1 (ISO 3)
Disposable
Single-use
Filter Efficiency
89.3% (PFE), 94.1% (BFE)
Includes
Knitted Earloops
Latex Free
Yes
Model
62470
Pore Size
0.1 μm (PFE), 3 μm (BFE)
Series
Kimtech™ M3 Sterile