▸
Microscope Slides
▸
Microscope Slide Storage
▸
Microscopy Slide Holders
▸
Microscope Slide Mailer
Actual product may vary
Microscope Slide Mailer
Allows for the safe storage and transport of microscope slides.
Have Questions?
Change view
Catalog Number
Slide Capacity
KLC62103
5
KLC62102
2
2 Options
Catalog number
KLC62103
Price (SGD)
23.00
Special offer
Ends:
31-Dec-2026
282.00
Save 259.00 (92%)
Box
Add to cart
Slide Capacity:
5
Price (SGD)
23.00
Special offer
Ends:
31-Dec-2026
282.00
Save 259.00 (92%)
Box
Add to cart
These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
Available in four sizes.
These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
Available in four sizes.
Specifications
Type
Slide Mailer
Slide Capacity
5
For Use With (Equipment)
Microscope Slides
Unit Size
Box