▸
Filters and Filtration
▸
Filter Plates
▸
Series 16 3 Porosity Biplane Filter Disc
Series 16 3 Porosity Biplane Filter Disc
Have Questions?
Catalog Number
Diameter (Metric)
KLC62003
200 mm
Catalog number
KLC62003
Price (SGD)
34.00
Special offer
Ends:
06-Sep-2026
409.00
Save 375.00 (92%)
Box
Add to cart
Diameter (Metric):
200 mm
Price (SGD)
34.00
Special offer
Ends:
06-Sep-2026
409.00
Save 375.00 (92%)
Box
Add to cart
Specifications
Quantity
1Pack
Type
Filter Disc
Height (Metric)
11.5 mm
Diameter (Metric)
200 mm
Unit Size
Box