Series 16 2 Porosity Biplane Filter Disc
Product Image
Series 16 2 Porosity Biplane Filter Disc
Have Questions?
Catalog NumberDiameter (Metric)
KLC62002200 mm
Catalog number KLC62002
Price (SGD)
21.00
Special offer
Ends: 31-Mar-2026
338.00
Save 317.00 (94%)
BX
Add to cart
Diameter (Metric):
200 mm
Price (SGD)
21.00
Special offer
Ends: 31-Mar-2026
338.00
Save 317.00 (94%)
BX
Add to cart
Specifications
Quantity1Pack
TypeFilter Disc
Height (Metric)11.5 mm
Diameter (Metric)200 mm
Unit SizeBX