▸
Filters and Filtration
▸
Filtration Membrane Discs
▸
Filter Disc, Surfaces Plane, Ground Cylindrical, 200 mm
Ansell
Filter Disc, Surfaces Plane, Ground Cylindrical, 200 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
Have Questions?
Catalog Number
Porosity
KLC62001
1
Catalog number
KLC62001
Price (SGD)
34.00
Special offer
Ends:
31-Dec-2026
409.00
Save 375.00 (92%)
Box
Add to cart
Porosity:
1
Price (SGD)
34.00
Special offer
Ends:
31-Dec-2026
409.00
Save 375.00 (92%)
Box
Add to cart
Highly resistant to most chemicals and temperatures up to 500°C
The biplane version may be required in specific installation circumstances
Specifications
Material
Borosilicate Glass (3.3)
Pore Size
1 μm
Sterility
Non-sterile
No. per Pack
1 Ea.
Porosity
1
Thickness (Metric)
11.5 mm ± 0.5 mm
Diameter (Metric)
200 mm
Unit Size
Box