G10 Blue Nitrile Gloves
G10 Blue Nitrile Gloves
Actual product may vary
G10 Blue Nitrile Gloves
G10 Blue Nitrile Gloves
G10 Blue Nitrile Gloves
G10 Blue Nitrile Gloves
KleenGuard™ G10 gloves provide durability and strength for light-duty tasks. Ideal for industrial, manufacturing and food processing applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC57374X-Large
KLC57371Small
KLC57372Medium
KLC57373Large
Catalog number KLC57374
Price (SGD)
-
Size:
X-Large
KleenGuard™ G10 gloves provide durability and strength for light-duty tasks. Ideal for industrial, manufacturing and food processing applications.
  • Powder free, nitrile
  • Textured fingertips provide enhanced grip and dexterity
  • Ambidextrous
  • Beaded cuff
  • Puncture-resistant formulation delivers barrier protection against chemical splash, dirt, grease and oil
  • Ideal for cleaning and maintenance, manufacturing, food processing, electronic assembly and painting applications
Specifications
Certifications/ComplianceUS FDA regulations and CFIA approved
Accelerator FreeYes
ApplicationsIndustrial
AQL1.5
ColorBlue
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)6 mil
Finger Thickness (Metric)0.15 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)4.3 mil
Palm Thickness (Metric)0.11 mm
PowderedPowder Free
Product TypeNitrile Gloves
SizeX-Large
SterilityNon-sterile
Thickness (English)6 mil
Thickness (Metric)0.11 mm
Model57374
Length (English)9.5 in.
Length (Metric)24 cm
Unit SizeCase