Hand-specific Kimtech™ Pure G3 white nitrile gloves.
Have Questions?
Change view
Catalog Number
Size
KLC56893
8.5
KLC56888
X-Small, 6
KLC56890
7
3 Options
Catalog number KLC56893
Price (SGD)
550.00
CS
Add to cart
Size:
8.5
Price (SGD)
550.00
CS
Add to cart
200 pairs of Kimtech™ Pure G3 white nitrile gloves. Hand-specific gloves.
Provides enhanced sterile cleanroom protection against a wide range of contaminants, chemicals, viruses, and cytotoxic drugs.
With an AQL of 1.5, the anti-static tested, latex and silicon free gloves feature a textured finish for improved grip and sensitivity while being suitable for double donning.
Validated to a Sterility Assurance Level (SAL) of 10-6, performing with a cleanliness level of a maximum of 950 particles >0.5μm/cm2, contains low endotoxins, meets FDA food contact and USP 800 guidelines.
The hand-specific gloves are cleanroom-ready; double-packaged in polyethylene bags to ensure your processes and workflows can continue operating efficiently within ISO 3 level cleanrooms and higher including Biotech, Aseptic processing, Parenteral drug and Ophthalmic manufacturing environments.